Learn More
Waters Corp Waters Oasis MCX 96-well Plate, 10 mg sorbent per well, 30 μm, 1/pk
Catalog No. 50780153 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MCX 96-well plate features a novel, mixed-mode polymeric sorbent for selective extraction of basic compounds. Its unique balance of hydrophobicity and water-wettability ensures consistent results, even if wells dry out. It is compatible with various liquid handling systems for high-throughput applications.
  • Novel, mixed-mode polymeric sorbent for basic compounds with cation-exchange groups.
  • Dual retention modes: Reversed-phase and cation exchange.
  • Ion exchange capacity: 1 meq/gram.
  • Particle size: 30 μm; Sorbent weight: 10 mg per well.
  • Stable across a wide pH range (pH 0 to 14).
  • Greater binding capacity than silica-based mixed-mode SPE sorbents.
  • 96-well plate format, meeting ANSI guidelines.
  • Innovative two-stage well design for high throughput and recovery.
  • Polypropylene construction; mass spectrometry compatible.
  • Water wettable.
Catalog No. 50-780-153 Supplier Waters Corp Supplier No. 186000259
The Oasis MCX 96-well plate features a novel, mixed-mode polymeric sorbent for selective extraction of basic compounds. Its unique balance of hydrophobicity and water-wettability ensures consistent results, even if wells dry out. It is compatible with various liquid handling systems for high-throughput applications.
  • Novel, mixed-mode polymeric sorbent for basic compounds with cation-exchange groups.
  • Dual retention modes: Reversed-phase and cation exchange.
  • Ion exchange capacity: 1 meq/gram.
  • Particle size: 30 μm; Sorbent weight: 10 mg per well.
  • Stable across a wide pH range (pH 0 to 14).
  • Greater binding capacity than silica-based mixed-mode SPE sorbents.
  • 96-well plate format, meeting ANSI guidelines.
  • Innovative two-stage well design for high throughput and recovery.
  • Polypropylene construction; mass spectrometry compatible.
  • Water wettable.