Learn More
Waters Corp Oasis MCX 96-well Plate, 30 mg Sorbent per Well, 30 µm, 1/pk
Catalog No. 50818655 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MCX 96-well plate contains the Oasis MCX sorbent, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups. The unique balance of hydrophobicity and water-wettability of the Oasis MCX sorbent means you will never have to worry about poor results if individual wells of the 96-well plate dry out during the critical steps prior to sample loading. The Oasis MCX 96-well plate is designed to be used on many manifold configurations and most robotic liquid handling systems.
Catalog No. 50-818-655 Supplier Waters Corp Supplier No. 186000248
The Oasis MCX 96-well plate contains the Oasis MCX sorbent, which is a novel, mixed-mode polymeric sorbent that has been optimized to achieve higher selectivity and sensitivity for extracting basic compounds with cation-exchange groups. The unique balance of hydrophobicity and water-wettability of the Oasis MCX sorbent means you will never have to worry about poor results if individual wells of the 96-well plate dry out during the critical steps prior to sample loading. The Oasis MCX 96-well plate is designed to be used on many manifold configurations and most robotic liquid handling systems.