Learn More
Waters Corp Oasis 96-well Plate, MCX, 30 mg Sorbent per Well, 60 μm
Catalog No. 50780147 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis MCX 96-well plate contains a novel, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups. Its unique balance of hydrophobicity and water-wettability ensures good results even if individual wells dry out during critical steps before sample loading.
  • Optimized for selective extraction of basic compounds with cation-exchange groups.
  • Features a 60 μm particle size, recommended for viscous samples.
  • Compatible with many manifold configurations and robotic liquid handling systems.
  • Constructed from polypropylene for chemical stability and inertness.
  • Offers a broad operational pH range of 0 to 14.
  • Provides 30 mg sorbent weight per well across 96 wells.
  • Ion exchange capacity of 1 meq/gram and a pore size of 80 Å.
  • Water wettable and mass spec compatible.
Catalog No. 50-780-147 Supplier Waters Corp Supplier No. 186000250
The Oasis MCX 96-well plate contains a novel, mixed-mode polymeric sorbent optimized for higher selectivity and sensitivity in extracting basic compounds with cation-exchange groups. Its unique balance of hydrophobicity and water-wettability ensures good results even if individual wells dry out during critical steps before sample loading.
  • Optimized for selective extraction of basic compounds with cation-exchange groups.
  • Features a 60 μm particle size, recommended for viscous samples.
  • Compatible with many manifold configurations and robotic liquid handling systems.
  • Constructed from polypropylene for chemical stability and inertness.
  • Offers a broad operational pH range of 0 to 14.
  • Provides 30 mg sorbent weight per well across 96 wells.
  • Ion exchange capacity of 1 meq/gram and a pore size of 80 Å.
  • Water wettable and mass spec compatible.