Learn More
Waters Corp Waters Oasis MCX Plus Short Cartridge, 225 mg, 60 μm, 50/pk
Catalog No. 50781816 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis MCX Plus cartridges contain a novel, mixed-mode polymeric sorbent, optimized for higher selectivity and sensitivity in extracting basic compounds. The sorbent is water wettable, maintaining high retention and recoveries even if it runs dry.
  • Contains mixed-mode polymeric sorbent for higher selectivity and sensitivity in extracting basic compounds.
  • Water wettable sorbent maintains high retention and excellent recoveries even if it runs dry.
  • 60 μm particle size is recommended for viscous samples.
  • Easily attaches to pumps and syringes and can be fitted with removable reservoirs for various flow devices.
  • Offers high SPE recovery, clean extracts, low matrix effects, and low method variability.
  • Enables fast, straightforward method development with dual modes of retention (ion-exchange and reversed-phase) and is stable from pH 0 to 14.
Catalog No. 50-781-816 Supplier Waters Corp Supplier No. 186003516
Oasis MCX Plus cartridges contain a novel, mixed-mode polymeric sorbent, optimized for higher selectivity and sensitivity in extracting basic compounds. The sorbent is water wettable, maintaining high retention and recoveries even if it runs dry.
  • Contains mixed-mode polymeric sorbent for higher selectivity and sensitivity in extracting basic compounds.
  • Water wettable sorbent maintains high retention and excellent recoveries even if it runs dry.
  • 60 μm particle size is recommended for viscous samples.
  • Easily attaches to pumps and syringes and can be fitted with removable reservoirs for various flow devices.
  • Offers high SPE recovery, clean extracts, low matrix effects, and low method variability.
  • Enables fast, straightforward method development with dual modes of retention (ion-exchange and reversed-phase) and is stable from pH 0 to 14.