Learn More
Waters Corp WATERS CORPORATION Oasis MCX Cartridge, 35 cc, 6 g Sorbent, 60 μm
Catalog No. 50819117 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis MCX cartridges are designed for solid-phase extraction, featuring a mixed-mode cation exchange sorbent for bases. They are built upon a unique water-wettable Oasis HLB Copolymer, providing both reversed-phase and ion exchange modes of retention for enhanced performance.
  • Functions as a mixed-mode cation exchange sorbent for bases.
  • Constructed with water-wettable Oasis HLB Copolymer.
  • Offers both reversed-phase and ion exchange retention capabilities.
  • Ensures greater cleanup selectivity and sensitivity for basic compounds.
Catalog No. 50-819-117 Supplier Waters Corp Supplier No. 186000778
Oasis MCX cartridges are designed for solid-phase extraction, featuring a mixed-mode cation exchange sorbent for bases. They are built upon a unique water-wettable Oasis HLB Copolymer, providing both reversed-phase and ion exchange modes of retention for enhanced performance.
  • Functions as a mixed-mode cation exchange sorbent for bases.
  • Constructed with water-wettable Oasis HLB Copolymer.
  • Offers both reversed-phase and ion exchange retention capabilities.
  • Ensures greater cleanup selectivity and sensitivity for basic compounds.