Learn More
Waters Corp Oasis Vac Cartridge, PRiME HLB, 1 cc, 30 mg Sorbent
Catalog No. NC1535587 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SPE sorbent simplifies protocols and provides reversed-phase cleanup of acidic, basic, and neutral compounds from complex sample matrices. It effectively removes over 95% of common matrix interferences like salts, proteins, and phospholipids. The water-wettable sorbent eliminates traditional conditioning and equilibration, speeding up processing and reducing solvent costs.
  • Provides reversed-phase cleanup of acidic, basic, and neutral compounds.
  • Removes over 95% of common matrix interferences (e.g., salts, proteins, phospholipids).
  • Eliminates conditioning and equilibration steps, simplifying sample preparation.
  • Facilitates faster processing of viscous samples; flow times are 30–50% faster for urine and plasma.
  • Optimized frit and sorbent design allows 4x faster processing with 1:1 diluted plasma.
  • Supports 3-step or 2-step cleanup processes depending on sample needs.
Catalog No. NC1535587 Supplier Waters Corp Supplier No. 186008055
This SPE sorbent simplifies protocols and provides reversed-phase cleanup of acidic, basic, and neutral compounds from complex sample matrices. It effectively removes over 95% of common matrix interferences like salts, proteins, and phospholipids. The water-wettable sorbent eliminates traditional conditioning and equilibration, speeding up processing and reducing solvent costs.
  • Provides reversed-phase cleanup of acidic, basic, and neutral compounds.
  • Removes over 95% of common matrix interferences (e.g., salts, proteins, phospholipids).
  • Eliminates conditioning and equilibration steps, simplifying sample preparation.
  • Facilitates faster processing of viscous samples; flow times are 30–50% faster for urine and plasma.
  • Optimized frit and sorbent design allows 4x faster processing with 1:1 diluted plasma.
  • Supports 3-step or 2-step cleanup processes depending on sample needs.