Learn More
Waters Corp Oasis Vac Cartridge, PRiME HLB, 3 cc, 60 mg Sorbent
Catalog No. 502038310 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis PRiME HLB sorbent provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices, simplifying solid phase extraction in routine analysis. It produces cleaner samples by removing over 95% of common matrix interferences like salts, proteins, and phospholipids. Viscous samples can be processed with faster flows and less plugging, and the water-wettable sorbent does not require conditioning and equilibration for excellent recoveries.
  • Stable at pH extremes (pH 1 to 12) and in a wide range of solvents.
  • Offers extraordinary retention of polar compounds.
  • Provides a relative hydrophobic retention capacity that is three times higher than traditional silica-based SPE sorbents.
Catalog No. 50-203-8310 Supplier Waters Corp Supplier No. 186008056
The Oasis PRiME HLB sorbent provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices, simplifying solid phase extraction in routine analysis. It produces cleaner samples by removing over 95% of common matrix interferences like salts, proteins, and phospholipids. Viscous samples can be processed with faster flows and less plugging, and the water-wettable sorbent does not require conditioning and equilibration for excellent recoveries.
  • Stable at pH extremes (pH 1 to 12) and in a wide range of solvents.
  • Offers extraordinary retention of polar compounds.
  • Provides a relative hydrophobic retention capacity that is three times higher than traditional silica-based SPE sorbents.