Learn More
Waters Corp Waters Solid Phase Extraction Vac Cartridge, PRiME HLB, 3 cc, 150 mg Sorbent, 100/pk
Catalog No. 501461923 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis PRiME HLB sets a new standard for solid phase extraction cleanup in routine analysis. It provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices, simplifying SPE. This sorbent produces cleaner samples by removing over >95% of common matrix interferences like salts, proteins, and phospholipids, and allows for faster processing of viscous samples with less plugging. The 3 cc cartridge with 150 mg sorbent enables greater sample cleanup via pass-thru protocol for enrichment.
  • Sets a new performance standard for solid phase extraction cleanup.
  • Provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex matrices, simplifying SPE protocols.
  • Removes over >95% of common matrix interferences (salts, proteins, phospholipids) and enables faster processing of viscous samples and greater sample cleanup capacity.
Catalog No. 50-146-1923 Supplier Waters Corp Supplier No. 186008717
Oasis PRiME HLB sets a new standard for solid phase extraction cleanup in routine analysis. It provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices, simplifying SPE. This sorbent produces cleaner samples by removing over >95% of common matrix interferences like salts, proteins, and phospholipids, and allows for faster processing of viscous samples with less plugging. The 3 cc cartridge with 150 mg sorbent enables greater sample cleanup via pass-thru protocol for enrichment.
  • Sets a new performance standard for solid phase extraction cleanup.
  • Provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex matrices, simplifying SPE protocols.
  • Removes over >95% of common matrix interferences (salts, proteins, phospholipids) and enables faster processing of viscous samples and greater sample cleanup capacity.