Learn More
Waters Corp Waters Vac Cartridge, Oasis PRiME HLB, 6 cc, 200 mg Sorbent
Catalog No. 502038311 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis PRiME HLB sets a new performance standard for solid phase extraction cleanup in routine analysis. It provides reversed-phase cleanup for acidic, basic, and neutral compounds, removing over 95% of common matrix interferences. The water-wettable copolymer eliminates conditioning and equilibration, speeding up processing and allowing higher capacity with less sorbent.
  • Simplifies solid phase extraction (SPE) with easy protocols.
  • Removes over 95% of matrix interferences: salts, proteins, and phospholipids.
  • Processes viscous samples with faster flows and reduced plugging.
  • No conditioning or equilibration required, saving processing time and solvent costs.
  • Macroporous copolymer with reversed-phase capability for enhanced capture of polar analytes and superior wettability.
  • 30 mg sorbent offers capacity equivalent to 100 mg of C18.
Catalog No. 50-203-8311 Supplier Waters Corp Supplier No. 186008057
Oasis PRiME HLB sets a new performance standard for solid phase extraction cleanup in routine analysis. It provides reversed-phase cleanup for acidic, basic, and neutral compounds, removing over 95% of common matrix interferences. The water-wettable copolymer eliminates conditioning and equilibration, speeding up processing and allowing higher capacity with less sorbent.
  • Simplifies solid phase extraction (SPE) with easy protocols.
  • Removes over 95% of matrix interferences: salts, proteins, and phospholipids.
  • Processes viscous samples with faster flows and reduced plugging.
  • No conditioning or equilibration required, saving processing time and solvent costs.
  • Macroporous copolymer with reversed-phase capability for enhanced capture of polar analytes and superior wettability.
  • 30 mg sorbent offers capacity equivalent to 100 mg of C18.