Learn More
Waters Corp Waters Oasis PRiME HLB Vac Cartridge, 6 cc, 500 mg, Reversed Phase
Catalog No. 501461924 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis PRiME HLB is a reversed–phase sorbent designed for solid phase extraction cleanup in routine analysis. It simplifies SPE protocols, producing cleaner samples by removing over 95% of common matrix interferences like salts, proteins, and phospholipids. It also allows for faster processing of viscous samples with less plugging.
  • Does not require conditioning and equilibration to provide excellent recoveries.
  • Offers a neutral polar hook for enhanced retention of polar analytes by providing reversed–phase capacity.
  • Delivers stability at pH extremes and in a wide range of solvents.
  • Provides extraordinary retention of polar compounds.
  • Has a relative hydrophobic retention capacity that is 3 times higher than traditional silica–based SPE sorbents like C18.
Catalog No. 50-146-1924 Supplier Waters Corp Supplier No. 186008718
The Oasis PRiME HLB is a reversed–phase sorbent designed for solid phase extraction cleanup in routine analysis. It simplifies SPE protocols, producing cleaner samples by removing over 95% of common matrix interferences like salts, proteins, and phospholipids. It also allows for faster processing of viscous samples with less plugging.
  • Does not require conditioning and equilibration to provide excellent recoveries.
  • Offers a neutral polar hook for enhanced retention of polar analytes by providing reversed–phase capacity.
  • Delivers stability at pH extremes and in a wide range of solvents.
  • Provides extraordinary retention of polar compounds.
  • Has a relative hydrophobic retention capacity that is 3 times higher than traditional silica–based SPE sorbents like C18.