Learn More
Waters Corp Waters 96-well Plate, PRiME HLB, 10 mg Sorbent per Well
Catalog No. 502038309 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis PRiME HLB sets a new standard for solid phase extraction cleanup, providing reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices. It simplifies SPE with easy-to-follow protocols, removing over 95% of common matrix interferences like salts, proteins, and phospholipids for cleaner samples.
  • Simplified SPE protocols.
  • Removes over 95% of common matrix interferences (salts, proteins, phospholipids).
  • Faster processing for viscous samples due to optimized frit and sorbent.
  • Water-wettable sorbent, no conditioning or equilibration needed.
  • Versatile reversed-phase cleanup for acidic, basic, and neutral compounds.
  • Compatible with robotic systems for high-throughput SPE.
Catalog No. 50-203-8309 Supplier Waters Corp Supplier No. 186008053
Oasis PRiME HLB sets a new standard for solid phase extraction cleanup, providing reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices. It simplifies SPE with easy-to-follow protocols, removing over 95% of common matrix interferences like salts, proteins, and phospholipids for cleaner samples.
  • Simplified SPE protocols.
  • Removes over 95% of common matrix interferences (salts, proteins, phospholipids).
  • Faster processing for viscous samples due to optimized frit and sorbent.
  • Water-wettable sorbent, no conditioning or equilibration needed.
  • Versatile reversed-phase cleanup for acidic, basic, and neutral compounds.
  • Compatible with robotic systems for high-throughput SPE.