Learn More
Waters Corp Waters Corporation Oasis 96-Well Plate, PRiME HLB, 30 mg Sorbent
Catalog No. NC1109119 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis PRiME HLB sorbent sets a new performance standard for solid phase extraction cleanup. It delivers reversed-phase cleanup for acidic, basic, and neutral compounds, removing over 95% of common matrix interferences like salts, proteins, and phospholipids. Designed for simplified SPE, it allows faster processing of viscous samples with less plugging.
  • Features 96 wells, each containing 30 mg of sorbent.
  • Eliminates conditioning and equilibration steps.
  • Optimized frit and sorbent design allows for easier flow of viscous samples through cartridges and plates.
  • Achieves faster flow rates compared to common matrices such as plasma, urine, and milk.
  • Requires less vacuum or positive pressure for optimal load, wash, and elution flow rates.
  • Compatible with most liquid-handling robotic systems for automated high-throughput SPE.
  • Constructed from durable polypropylene material.
Catalog No. NC1109119 Supplier Waters Corp Supplier No. 186008054
Oasis PRiME HLB sorbent sets a new performance standard for solid phase extraction cleanup. It delivers reversed-phase cleanup for acidic, basic, and neutral compounds, removing over 95% of common matrix interferences like salts, proteins, and phospholipids. Designed for simplified SPE, it allows faster processing of viscous samples with less plugging.
  • Features 96 wells, each containing 30 mg of sorbent.
  • Eliminates conditioning and equilibration steps.
  • Optimized frit and sorbent design allows for easier flow of viscous samples through cartridges and plates.
  • Achieves faster flow rates compared to common matrices such as plasma, urine, and milk.
  • Requires less vacuum or positive pressure for optimal load, wash, and elution flow rates.
  • Compatible with most liquid-handling robotic systems for automated high-throughput SPE.
  • Constructed from durable polypropylene material.