Learn More
Waters Corp Waters Oasis PRiME HLB Plus Light Cartridge, 100 mg Sorbent, 50 pk
Catalog No. 501462015 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis PRiME HLB sets a new performance standard for solid phase extraction cleanup in routine analysis. This sorbent provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices. It simplifies solid phase extraction with easy-to-follow protocols and produces cleaner samples by removing over 95% of common matrix interferences, including salts, proteins, and phospholipids. Its extreme water-wettability eliminates conditioning and equilibration steps, saving time and reducing solvent costs.
  • Provides reversed-phase cleanup of acidic, basic, and neutral compounds from complex sample matrices.
  • Produces cleaner samples by removing greater than 95% of common matrix interferences.
  • Simplifies solid phase extraction with easy-to-follow protocols.
Catalog No. 50-146-2015 Supplier Waters Corp Supplier No. 186008886
Oasis PRiME HLB sets a new performance standard for solid phase extraction cleanup in routine analysis. This sorbent provides reversed-phase cleanup for acidic, basic, and neutral compounds from complex sample matrices. It simplifies solid phase extraction with easy-to-follow protocols and produces cleaner samples by removing over 95% of common matrix interferences, including salts, proteins, and phospholipids. Its extreme water-wettability eliminates conditioning and equilibration steps, saving time and reducing solvent costs.
  • Provides reversed-phase cleanup of acidic, basic, and neutral compounds from complex sample matrices.
  • Produces cleaner samples by removing greater than 95% of common matrix interferences.
  • Simplifies solid phase extraction with easy-to-follow protocols.