Learn More
Waters Corp Oasis PRiME HLB Plus Short Cartridge, 335 mg Sorbent per Cartridge, 50/pkg;Reversed-phase, Water Wettable, Mass Spec Compatible
Catalog No. 501462016 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis PRiME HLB Plus Short Cartridge, 335 mg Sorbent per Cartridge, 50/pkg;Reversed-phase, Water Wettable, Mass Spec Compatible
Catalog No. 50-146-2016 Supplier Waters Corp Supplier No. 186008887
Oasis PRiME HLB Plus Short Cartridge, 335 mg Sorbent per Cartridge, 50/pkg;Reversed-phase, Water Wettable, Mass Spec Compatible