Learn More
Waters Corp Waters μElution Plate, PRiME MCX, 96-well, 2 mg Sorbent, 30 μm
Catalog No. 501528583 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis PRiME MCX is a mixed-mode polymeric sorbent designed for extracting basic compounds with cation-exchange groups. It is optimized for higher selectivity and sensitivity, ensuring consistent results even if wells dry out before sample loading. This product simplifies Solid Phase Extraction (SPE) with fast, easy protocols and produces cleaner samples by removing over 99% of phospholipids.
  • Mixed-mode polymeric sorbent for extracting basic compounds
  • Optimized for higher selectivity and sensitivity
  • 96-well plate format
  • 2 mg sorbent per well
  • 30 μm particle size
  • Water-wettable sorbent
  • Constructed from polypropylene
  • Mass spectrometer compatible
  • Simplifies Solid Phase Extraction (SPE) with fast, easy protocols
  • Removes over 99% of phospholipids for cleaner samples
  • Does not require conditioning and equilibration
Catalog No. 50-152-8583 Supplier Waters Corp Supplier No. 186008914
Oasis PRiME MCX is a mixed-mode polymeric sorbent designed for extracting basic compounds with cation-exchange groups. It is optimized for higher selectivity and sensitivity, ensuring consistent results even if wells dry out before sample loading. This product simplifies Solid Phase Extraction (SPE) with fast, easy protocols and produces cleaner samples by removing over 99% of phospholipids.
  • Mixed-mode polymeric sorbent for extracting basic compounds
  • Optimized for higher selectivity and sensitivity
  • 96-well plate format
  • 2 mg sorbent per well
  • 30 μm particle size
  • Water-wettable sorbent
  • Constructed from polypropylene
  • Mass spectrometer compatible
  • Simplifies Solid Phase Extraction (SPE) with fast, easy protocols
  • Removes over 99% of phospholipids for cleaner samples
  • Does not require conditioning and equilibration