Learn More
Waters Corp Waters Corporation 96-well Plate, Oasis PRiME MCX, 30 mg, 30 μm
Catalog No. 501528585 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis PRiME MCX 96-well plate uses a mixed-mode polymeric sorbent for solid-phase extraction. It ensures robust performance by maintaining water-wettability even if wells dry out. This highly efficient, reversed-phase and ion-exchange product effectively isolates basic compounds from complex matrices. Made of polypropylene, it offers chemical stability, enhancing chromatographic analysis with improved resolution, sensitivity, and throughput.
  • Contains a novel, mixed-mode polymeric sorbent.
  • Unique water-wettability prevents poor results if wells dry out.
  • Designed to capture and concentrate basic compounds, removing matrix interferences.
  • Polypropylene construction offers chemical stability and inertness.
  • Improved resolution, sensitivity, and throughput.
Catalog No. 50-152-8585 Supplier Waters Corp Supplier No. 186008916
The Oasis PRiME MCX 96-well plate uses a mixed-mode polymeric sorbent for solid-phase extraction. It ensures robust performance by maintaining water-wettability even if wells dry out. This highly efficient, reversed-phase and ion-exchange product effectively isolates basic compounds from complex matrices. Made of polypropylene, it offers chemical stability, enhancing chromatographic analysis with improved resolution, sensitivity, and throughput.
  • Contains a novel, mixed-mode polymeric sorbent.
  • Unique water-wettability prevents poor results if wells dry out.
  • Designed to capture and concentrate basic compounds, removing matrix interferences.
  • Polypropylene construction offers chemical stability and inertness.
  • Improved resolution, sensitivity, and throughput.