Learn More
Waters Corp Solid Phase Extraction Plate, 96-Well, 10 mg Sorbent, 30 μm Particle
Catalog No. 502391857 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well plate uses a mixed-mode polymeric sorbent, balancing hydrophobicity and water-wettability to prevent drying during pre-sample loading. It offers efficient reversed-phase and ion-exchange solid-phase extraction for capturing basic chemicals and removing matrix interferences.
  • Mixed-mode polymeric sorbent with balanced hydrophobicity and water-wettability.
  • Prevents well drying during critical pre-sample loading.
  • Efficient reversed-phase and ion-exchange solid-phase extraction.
  • Captures basic chemicals, removes matrix interferences.
  • Controlled ion-exchange capacity; no silanol groups.
  • Constructed from chemically stable, inert polypropylene.
  • Enhances resolution, sensitivity, throughput, and analysis efficiency.
  • Supports wide range of flow rates and back pressures.
  • Reduces time and cost per sample; improves results quality.
Catalog No. 50-239-1857 Supplier Waters Corp Supplier No. 186008915
Add to Cart
Add to Cart
This 96-well plate uses a mixed-mode polymeric sorbent, balancing hydrophobicity and water-wettability to prevent drying during pre-sample loading. It offers efficient reversed-phase and ion-exchange solid-phase extraction for capturing basic chemicals and removing matrix interferences.
  • Mixed-mode polymeric sorbent with balanced hydrophobicity and water-wettability.
  • Prevents well drying during critical pre-sample loading.
  • Efficient reversed-phase and ion-exchange solid-phase extraction.
  • Captures basic chemicals, removes matrix interferences.
  • Controlled ion-exchange capacity; no silanol groups.
  • Constructed from chemically stable, inert polypropylene.
  • Enhances resolution, sensitivity, throughput, and analysis efficiency.
  • Supports wide range of flow rates and back pressures.
  • Reduces time and cost per sample; improves results quality.