Learn More
Waters Corp Oasis Vac Cartridge, PRiME MCX, 3 cc, 60 mg, 30 μm
Catalog No. 501528587 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis PRiME MCX 3 cc Vac Cartridge is a water-wettable, polymeric SPE sorbent designed to overcome limitations of traditional silica-based mixed-mode sorbents. It provides cleaner samples with reduced effort and minimizes matrix effects.
  • Minimizes matrix effects and chromatographic complexity; allows increased sample concentration.
  • Possesses significantly greater binding capacity than most silica-based mixed-mode SPE sorbents.
  • Offers exceptional pH stability for method development.
  • Provides high selectivity and sensitivity for basic compounds.
  • Ensures reproducible performance with 1.0 meq/g sulfonic acid ion exchange capacity.
  • Facilitates easier flow of viscous samples with optimized frit and sorbent designs.
  • Eliminates conditioning and equilibration steps for faster processing and reproducible cleanup.
Catalog No. 50-152-8587 Supplier Waters Corp Supplier No. 186008918
The Oasis PRiME MCX 3 cc Vac Cartridge is a water-wettable, polymeric SPE sorbent designed to overcome limitations of traditional silica-based mixed-mode sorbents. It provides cleaner samples with reduced effort and minimizes matrix effects.
  • Minimizes matrix effects and chromatographic complexity; allows increased sample concentration.
  • Possesses significantly greater binding capacity than most silica-based mixed-mode SPE sorbents.
  • Offers exceptional pH stability for method development.
  • Provides high selectivity and sensitivity for basic compounds.
  • Ensures reproducible performance with 1.0 meq/g sulfonic acid ion exchange capacity.
  • Facilitates easier flow of viscous samples with optimized frit and sorbent designs.
  • Eliminates conditioning and equilibration steps for faster processing and reproducible cleanup.