Learn More
Waters Corp Waters Oasis μElution Plate, 96 well, 2 mg, 30 μm, Peptide Method Development
Catalog No. 50466042 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis Peptide Method Development μElution Plate simplifies sample preparation for therapeutic peptide analysis in plasma. This 96-well plate features six rows each of two mixed-mode, ion-exchange chemistries, allowing rapid and easy method development using a single Solid Phase Extraction (SPE) protocol.
  • Streamlines sample preparation for enhanced throughput.
  • Efficient peptide recoveries from small samples (2 mg sorbent, 30 μm particle size).
  • μElution format: high-concentration eluates, no evaporation/reconstitution.
  • Direct injection compatibility with LC/MS systems.
  • Supports automated and manual high-throughput workflows.
  • Versatile for proteomics, biomarker discovery, and pharmacokinetic studies.
  • Quality control ensures consistent performance.
Catalog No. 50-466-042 Supplier Waters Corp Supplier No. 186004713
The Oasis Peptide Method Development μElution Plate simplifies sample preparation for therapeutic peptide analysis in plasma. This 96-well plate features six rows each of two mixed-mode, ion-exchange chemistries, allowing rapid and easy method development using a single Solid Phase Extraction (SPE) protocol.
  • Streamlines sample preparation for enhanced throughput.
  • Efficient peptide recoveries from small samples (2 mg sorbent, 30 μm particle size).
  • μElution format: high-concentration eluates, no evaporation/reconstitution.
  • Direct injection compatibility with LC/MS systems.
  • Supports automated and manual high-throughput workflows.
  • Versatile for proteomics, biomarker discovery, and pharmacokinetic studies.
  • Quality control ensures consistent performance.