Learn More
Med Vet International Oasis Skin Stapler, Sterile, 35 Wide, Each
Catalog No. NC1917644
Encompass_Preferred
This skin stapler is affordable and simple. It's simplistic design and quality construction provides consistent and reliable performance. It fits comfortably in your hand to enhance control of skin closure and also reduces surgery time. 35 staples preloaded Individually sterile Lever action staplers are the most popular design on the market Distinctive staple shuttling feature greatly reduces track friction, a primary cause of jamming Short trigger stroke for ease of use and nonstop feed Handle design comfortably fits a wide range of hand sizes Angled head allows visibility of incision to ensure precise staple placement during operation Arrow indicator assists in accurate staple alignment Staple count indicator allows you to view staples remaining to help minimize waste Quality construction and simple design assure reliable consistent performance Staple lines are more likely to prevent leakage of blood or air Regular staples measure 5.4mm x 3.6mm; Wide Staples measure 7mm x 4mm.
This item is not returnable. View return policy
Catalog No. NC1917644 Supplier Med Vet International Supplier No. 35W
 Restricted Item
Add to Cart
Add to Cart
This skin stapler is affordable and simple. It's simplistic design and quality construction provides consistent and reliable performance. It fits comfortably in your hand to enhance control of skin closure and also reduces surgery time. 35 staples preloaded Individually sterile Lever action staplers are the most popular design on the market Distinctive staple shuttling feature greatly reduces track friction, a primary cause of jamming Short trigger stroke for ease of use and nonstop feed Handle design comfortably fits a wide range of hand sizes Angled head allows visibility of incision to ensure precise staple placement during operation Arrow indicator assists in accurate staple alignment Staple count indicator allows you to view staples remaining to help minimize waste Quality construction and simple design assure reliable consistent performance Staple lines are more likely to prevent leakage of blood or air Regular staples measure 5.4mm x 3.6mm; Wide Staples measure 7mm x 4mm.