Learn More
Waters Corp Waters Oasis Method Development Sorbent Selection Kit, 1 cc, 30 mg, 30 μm
Catalog No. 50781795 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sorbent selection cartridge kit is designed for rapid development of Solid Phase Extraction (SPE) methods for LC/MS analysis. It offers both reversed-phase and ion exchange modes of retention for cleanup selectivity and sensitivity for acidic and/or basic compounds. The kit contains 40 cartridges, with 10 each of MAX, WAX, MCX, and WCX mixed-mode, ion-exchange chemistries.
  • Rapidly develop SPE methods for LC/MS analysis.
  • Includes 40 cartridges (10 each of MAX, WAX, MCX, WCX).
  • MAX is a strong anion exchanger for acids.
  • WAX is a weak anion exchanger for strong acidic compounds.
  • MCX is a strong cation exchanger for bases.
  • WCX is a weak cation exchanger for strong bases and quaternary amines.
  • Water wettable sorbents maintain retention and recovery even if they run dry.
  • No need for extra safety measures to prevent sorbent beds from drying out.
Catalog No. 50-781-795 Supplier Waters Corp Supplier No. 186003463
This sorbent selection cartridge kit is designed for rapid development of Solid Phase Extraction (SPE) methods for LC/MS analysis. It offers both reversed-phase and ion exchange modes of retention for cleanup selectivity and sensitivity for acidic and/or basic compounds. The kit contains 40 cartridges, with 10 each of MAX, WAX, MCX, and WCX mixed-mode, ion-exchange chemistries.
  • Rapidly develop SPE methods for LC/MS analysis.
  • Includes 40 cartridges (10 each of MAX, WAX, MCX, WCX).
  • MAX is a strong anion exchanger for acids.
  • WAX is a weak anion exchanger for strong acidic compounds.
  • MCX is a strong cation exchanger for bases.
  • WCX is a weak cation exchanger for strong bases and quaternary amines.
  • Water wettable sorbents maintain retention and recovery even if they run dry.
  • No need for extra safety measures to prevent sorbent beds from drying out.