Learn More
Waters Corp Waters Oasis Sorbent Selection Flangeless Cartridge Kit, 10 mg/1 cc, 30 μm, 40 pk
Catalog No. 50201609 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for the rapid development of solid phase extraction (SPE) methods for LC/MS analysis. It contains 40 flangeless cartridges, featuring 10 each of four mixed-mode, ion-exchange chemistries: MAX, WAX, MCX, and WCX. This enables quick and easy sample preparation method development using only two SPE protocols.
  • Rapid development of SPE methods for LC/MS analysis.
  • Contains 40 flangeless cartridges, with 10 each of MAX, WAX, MCX, and WCX sorbents.
  • Facilitates convenient method development, allowing selection among different sorbents.
  • Suitable for isolating unknown analytes, zwitterionic compounds, or mixtures of analytes with different retention/elution properties.
  • MCX chemistry for bases.
  • MAX chemistry for acids.
  • WCX chemistry for strong bases and quaternary amines.
  • WAX chemistry for strong acids.
Catalog No. 50-201-609 Supplier Waters Corp Supplier No. 186006344
This kit is designed for the rapid development of solid phase extraction (SPE) methods for LC/MS analysis. It contains 40 flangeless cartridges, featuring 10 each of four mixed-mode, ion-exchange chemistries: MAX, WAX, MCX, and WCX. This enables quick and easy sample preparation method development using only two SPE protocols.
  • Rapid development of SPE methods for LC/MS analysis.
  • Contains 40 flangeless cartridges, with 10 each of MAX, WAX, MCX, and WCX sorbents.
  • Facilitates convenient method development, allowing selection among different sorbents.
  • Suitable for isolating unknown analytes, zwitterionic compounds, or mixtures of analytes with different retention/elution properties.
  • MCX chemistry for bases.
  • MAX chemistry for acids.
  • WCX chemistry for strong bases and quaternary amines.
  • WAX chemistry for strong acids.