Learn More
Waters Corp Waters Sorbent Selection Plate, 10 mg Sorbent per Well, 30 μm, 96-well
Catalog No. 50781626 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Oasis sorbent selection plate simplifies solid phase extraction (SPE) method development. It features a 96-well format with three rows each of four distinct mixed-mode, ion-exchange chemistries (MAX, WAX, MCX, WCX). This comprehensive design helps identify optimal methods for isolating unknown analytes, zwitterionic compounds, or complex mixtures with diverse retention and elution characteristics, allowing for rapid sample preparation using just two SPE protocols.
  • Particle size: 30 μm.
  • Pore size: 80 Å.
  • Sorbent weight: 10 mg.
  • pH range: 0 to 14.
  • Material: Polypropylene.
  • Mass spectrometry compatible.
  • Water wettable.
  • Endcapped: No.
  • Sorbent substrate: Copolymer.
Catalog No. 50-781-626 Supplier Waters Corp Supplier No. 186003249
This Oasis sorbent selection plate simplifies solid phase extraction (SPE) method development. It features a 96-well format with three rows each of four distinct mixed-mode, ion-exchange chemistries (MAX, WAX, MCX, WCX). This comprehensive design helps identify optimal methods for isolating unknown analytes, zwitterionic compounds, or complex mixtures with diverse retention and elution characteristics, allowing for rapid sample preparation using just two SPE protocols.
  • Particle size: 30 μm.
  • Pore size: 80 Å.
  • Sorbent weight: 10 mg.
  • pH range: 0 to 14.
  • Material: Polypropylene.
  • Mass spectrometry compatible.
  • Water wettable.
  • Endcapped: No.
  • Sorbent substrate: Copolymer.