Learn More
Waters Corp Waters Corporation Oasis μElution Plate, Method Development, 96-well, 30 μm
Catalog No. 50782442 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This μElution Plate simplifies solid-phase extraction (SPE) method development. It features a patented 96-well format with three rows each of four mixed-mode, ion-exchange chemistries, enabling quick, high-throughput sample preparation compatible with robotic systems.
  • Simplifies SPE method development.
  • Patented 96-well format with mixed-mode ion-exchange chemistries.
  • Stable at pH extremes and in a wide range of solvents.
  • Extraordinary retention of polar compounds.
  • Three times higher hydrophobic retention capacity than traditional silica-based SPE sorbents.
  • Manufactured under ISO 9000 and cGMP standards.
  • Water wettable and mass spec compatible.
Catalog No. 50-782-442 Supplier Waters Corp Supplier No. 186004475
This μElution Plate simplifies solid-phase extraction (SPE) method development. It features a patented 96-well format with three rows each of four mixed-mode, ion-exchange chemistries, enabling quick, high-throughput sample preparation compatible with robotic systems.
  • Simplifies SPE method development.
  • Patented 96-well format with mixed-mode ion-exchange chemistries.
  • Stable at pH extremes and in a wide range of solvents.
  • Extraordinary retention of polar compounds.
  • Three times higher hydrophobic retention capacity than traditional silica-based SPE sorbents.
  • Manufactured under ISO 9000 and cGMP standards.
  • Water wettable and mass spec compatible.