Learn More
Waters Corp Waters Solid Phase Extraction Cartridge, WAX Chemistry, Mixed Mode, 60 μm, 225 mg
Catalog No. 50179320 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WAX Plus Short Cartridge features a polymeric weak anion-exchange, reversed-phase, mixed-mode sorbent. It is designed for highly selective sample preparation of strong acidic chemicals, offering a wide pH working range of 0 to 14. The sorbent is water-wettable, ensuring excellent recovery and retention even if it runs dry, thus eliminating the need for special measures before sample loading.
  • Polymeric weak anion-exchange, reversed-phase, mixed-mode sorbent.
  • Highly selective sample preparation for strong acidic chemicals.
  • Wide pH working range of 0 to 14.
  • Water-wettable for better recovery and higher retention, even when sorbent runs out.
  • No special measures required to prevent sorbent beds from drying out.
  • Includes benefits of the HLB sorbent.
  • Unique, carefully monitored production method.
  • 60 μm particle size is advised for viscous samples.
Catalog No. 50-179-320 Supplier Waters Corp Supplier No. 186003519
The Oasis WAX Plus Short Cartridge features a polymeric weak anion-exchange, reversed-phase, mixed-mode sorbent. It is designed for highly selective sample preparation of strong acidic chemicals, offering a wide pH working range of 0 to 14. The sorbent is water-wettable, ensuring excellent recovery and retention even if it runs dry, thus eliminating the need for special measures before sample loading.
  • Polymeric weak anion-exchange, reversed-phase, mixed-mode sorbent.
  • Highly selective sample preparation for strong acidic chemicals.
  • Wide pH working range of 0 to 14.
  • Water-wettable for better recovery and higher retention, even when sorbent runs out.
  • No special measures required to prevent sorbent beds from drying out.
  • Includes benefits of the HLB sorbent.
  • Unique, carefully monitored production method.
  • 60 μm particle size is advised for viscous samples.