Learn More
Waters Corp Waters Oasis WAX Vac Cartridge, 1 cc, 10 mg Sorbent, 30 μm
Catalog No. 50201496 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This reversed-phase, weak anion exchange mixed-mode sorbent is designed for fast, simple, and highly selective sample preparation of strong acidic compounds. The sorbent is water wettable, maintaining its capability for higher retention and excellent recoveries even if it runs dry, eliminating the need to take extraordinary steps to prevent drying out during critical steps prior to sample loading. A flangeless configuration allows for close packing of cartridges, facilitating processing in specialized automated SPE systems.
  • Sorbent maintains high retention and excellent recoveries even if it runs dry.
  • Reversed-phase, weak anion exchange mixed-mode sorbent.
  • Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds.
  • Flangeless configuration allows close packing for processing in specialized automated SPE systems.
Catalog No. 50-201-496 Supplier Waters Corp Supplier No. 186004651
This reversed-phase, weak anion exchange mixed-mode sorbent is designed for fast, simple, and highly selective sample preparation of strong acidic compounds. The sorbent is water wettable, maintaining its capability for higher retention and excellent recoveries even if it runs dry, eliminating the need to take extraordinary steps to prevent drying out during critical steps prior to sample loading. A flangeless configuration allows for close packing of cartridges, facilitating processing in specialized automated SPE systems.
  • Sorbent maintains high retention and excellent recoveries even if it runs dry.
  • Reversed-phase, weak anion exchange mixed-mode sorbent.
  • Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds.
  • Flangeless configuration allows close packing for processing in specialized automated SPE systems.