Learn More
Waters Corp Waters Vac Cartridge, Oasis WAX, 3 cc, 60 mg Sorbent, 30 μm
Catalog No. 50781147 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WAX 3 cc Vac Cartridge contains 60 mg of a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. It's optimized for fast, selective sample preparation of strong acidic compounds, remaining water wettable with excellent recoveries even if it runs dry.
  • Optimized for fast, selective sample preparation of strong acidic compounds.
  • Water wettable, ensuring retention and excellent recoveries even if dry.
  • Stable from pH 0 to 14 for wide polarity analyte analysis.
  • Delivers superior sample preparation and robust SPE methods.
  • High retention capacity improves recovery and reproducibility.
  • Expertly manufactured with stringent quality control for reproducibility.
  • Durable polypropylene construction with ultra-clean components.
  • Offers low-cost sample preparation.
Catalog No. 50-781-147 Supplier Waters Corp Supplier No. 186002490
The Oasis WAX 3 cc Vac Cartridge contains 60 mg of a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. It's optimized for fast, selective sample preparation of strong acidic compounds, remaining water wettable with excellent recoveries even if it runs dry.
  • Optimized for fast, selective sample preparation of strong acidic compounds.
  • Water wettable, ensuring retention and excellent recoveries even if dry.
  • Stable from pH 0 to 14 for wide polarity analyte analysis.
  • Delivers superior sample preparation and robust SPE methods.
  • High retention capacity improves recovery and reproducibility.
  • Expertly manufactured with stringent quality control for reproducibility.
  • Durable polypropylene construction with ultra-clean components.
  • Offers low-cost sample preparation.