Learn More
Waters Corp Waters Vac Cartridge, Oasis WAX, 6 cc, 500 mg, 60 μm
Catalog No. 50466019 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis WAX weak anion-exchange SPE chemistry offers fast, simple, and highly selective sample preparation and cleanup, utilizing a polymeric reversed-phase, weak ion exchange mixed-mode sorbent that is water wettable and compatible with MS friendly elution solvents.
  • Water-wettable polymeric sorbent allows direct loading of aqueous samples without conditioning.
  • Mixed-mode sorbent ensures highly selective sample preparation.
  • Superior sample preparation in strong acids, stable from pH 0 to 14.
  • High retention and recoveries, even if sorbent runs dry.
  • Reproducible delivery with 0.6 meq/g piperazine ion exchange capacity.
  • 6 cc vac cartridge with syringe barrel, suitable for automated SPE and vacuum manifolds.
  • Package includes 30 polypropylene cartridges.
Catalog No. 50-466-019 Supplier Waters Corp Supplier No. 186004647
Oasis WAX weak anion-exchange SPE chemistry offers fast, simple, and highly selective sample preparation and cleanup, utilizing a polymeric reversed-phase, weak ion exchange mixed-mode sorbent that is water wettable and compatible with MS friendly elution solvents.
  • Water-wettable polymeric sorbent allows direct loading of aqueous samples without conditioning.
  • Mixed-mode sorbent ensures highly selective sample preparation.
  • Superior sample preparation in strong acids, stable from pH 0 to 14.
  • High retention and recoveries, even if sorbent runs dry.
  • Reproducible delivery with 0.6 meq/g piperazine ion exchange capacity.
  • 6 cc vac cartridge with syringe barrel, suitable for automated SPE and vacuum manifolds.
  • Package includes 30 polypropylene cartridges.