Learn More
Waters Corp Waters Vac Cartridge, 1 cc, Flangeless, 30 mg/30 μm Sorbent
Catalog No. 50201645 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 1 cc flangeless cartridges contain a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. This sorbent is optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. It is water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry, eliminating the need to prevent sorbent beds from drying out before sample loading. The flangeless configuration allows for close packing of cartridges for processing in specialized automated SPE systems.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds
  • Water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry
  • Flangeless configuration for use in specialized automated SPE systems
Catalog No. 50-201-645 Supplier Waters Corp Supplier No. 186006500
The 1 cc flangeless cartridges contain a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. This sorbent is optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. It is water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry, eliminating the need to prevent sorbent beds from drying out before sample loading. The flangeless configuration allows for close packing of cartridges for processing in specialized automated SPE systems.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds
  • Water wettable, maintaining higher retention and excellent recoveries even if the sorbent runs dry
  • Flangeless configuration for use in specialized automated SPE systems