Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Vac Cartridge, PFAS Analysis, 6 cc, 150 mg Sorbent

Catalog No. 502391293 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Oasis WAX for PFAS Analysis is a mixed-mode, Weak Anion-eXchange, reversed-phase, water-wettable polymer designed for environmental testing laboratories to perform low-level quantification of per- and polyfluoroalkyl substances (PFAS) in various environmental samples, including drinking water, groundwater, surface water, wastewater, soil, and food, without the risk of cross-contamination.

  • Provides high purity, consistency, and quality for sample analysis.
  • Features a comprehensive quality control pre-screening for 32 PFAS.
  • Application-specific quality control testing minimizes false positives.
  • Mixed-mode, weak anion-exchange, reversed-phase chemistry.
  • Mass spectrometry compatible.
  • Particle size of 30 μm.
  • Sorbent weight of 150 mg.
  • pH range of 0 to 14.
  • Polypropylene construction, 6 cc barrel.

Catalog No. 50-239-1293 Supplier Waters Corp Supplier No. 186009344
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Oasis WAX for PFAS Analysis is a mixed-mode, Weak Anion-eXchange, reversed-phase, water-wettable polymer designed for environmental testing laboratories to perform low-level quantification of per- and polyfluoroalkyl substances (PFAS) in various environmental samples, including drinking water, groundwater, surface water, wastewater, soil, and food, without the risk of cross-contamination.

  • Provides high purity, consistency, and quality for sample analysis.
  • Features a comprehensive quality control pre-screening for 32 PFAS.
  • Application-specific quality control testing minimizes false positives.
  • Mixed-mode, weak anion-exchange, reversed-phase chemistry.
  • Mass spectrometry compatible.
  • Particle size of 30 μm.
  • Sorbent weight of 150 mg.
  • pH range of 0 to 14.
  • Polypropylene construction, 6 cc barrel.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.