Learn More
Waters Corp Vac Cartridge, WAX, 500 mg, 60 μm
Supplier: Waters Corp 186009346
This mixed-mode, Weak Anion-eXchange, reversed-phase, water-wettable polymer is designed for sample analysis in various matrices (drinking water, groundwater, surface water, wastewater, soil, food). It enables environmental testing laboratories to perform low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples, ensuring extraction and concentration without cross-contamination via comprehensive quality control pre-screening for 32 PFAS, reducing false positive results.
- Delivers unmatched purity, consistency, and quality for sample analysis.
- Designed specifically for environmental testing laboratories.
- Enables PFAS extraction and concentration without cross-contamination.
- Comprehensive quality control pre-screening test for 32 PFAS.
- Provides assurance in results through application-specific quality control testing.