Learn More
Waters Corp Waters OASIS WAX Vac Cartridge, PFAS Analysis, 6 cc
Catalog No. 502039104 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WAX for PFAS Analysis is a mixed-mode, Weak Anion-eXchange, reversed-phase, water-wettable polymer that offers unmatched purity, consistency, and quality for sample analysis in various complex matrices, including drinking water, ground water, surface water, wastewater, soil, and food. It is specifically engineered for environmental testing laboratories that require low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples.
  • Mixed-mode, Weak Anion-eXchange, reversed-phase, water-wettable polymer
  • Designed for low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples
  • Enables PFAS extraction and concentration without the risk of cross-contamination
  • Comprehensive quality control pre-screening test for 32 PFAS
  • Ideal for use with EPA Method 533
Catalog No. 50-203-9104 Supplier Waters Corp Supplier No. 186009568
The Oasis WAX for PFAS Analysis is a mixed-mode, Weak Anion-eXchange, reversed-phase, water-wettable polymer that offers unmatched purity, consistency, and quality for sample analysis in various complex matrices, including drinking water, ground water, surface water, wastewater, soil, and food. It is specifically engineered for environmental testing laboratories that require low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples.
  • Mixed-mode, Weak Anion-eXchange, reversed-phase, water-wettable polymer
  • Designed for low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental samples
  • Enables PFAS extraction and concentration without the risk of cross-contamination
  • Comprehensive quality control pre-screening test for 32 PFAS
  • Ideal for use with EPA Method 533