Learn More
Waters Corp Oasis WAX Vac Cartridge, PFAS Analysis, 6 cc, 500 mg, 60 μm
Catalog No. 502392352 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WAX for PFAS Analysis cartridge is a mixed-mode, Weak Anion-eXchange (WAX), reversed-phase, water-wettable polymer, designed for high-purity sample analysis.
  • Designed for low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental and complex matrices.
  • Ensures efficient PFAS extraction and concentration with reduced cross-contamination via comprehensive quality control pre-screening for 32 PFAS.
  • Features 0.6 meq/g piperazine-ion-exchange capacity and operates within a pH range of 0 to 14.
  • Sorbent maintains retention and recovery even when dry, simplifying sample loading.
  • Syringe-barrel cartridges are compatible with vacuum manifolds and automated SPE devices.
  • Ultra-clean design prevents contamination and analytical interference.
  • Particle size of 60 μm is recommended for viscous samples.
Catalog No. 50-239-2352 Supplier Waters Corp Supplier No. 186009347
The Oasis WAX for PFAS Analysis cartridge is a mixed-mode, Weak Anion-eXchange (WAX), reversed-phase, water-wettable polymer, designed for high-purity sample analysis.
  • Designed for low-level quantification of per- and polyfluoroalkyl substances (PFAS) in environmental and complex matrices.
  • Ensures efficient PFAS extraction and concentration with reduced cross-contamination via comprehensive quality control pre-screening for 32 PFAS.
  • Features 0.6 meq/g piperazine-ion-exchange capacity and operates within a pH range of 0 to 14.
  • Sorbent maintains retention and recovery even when dry, simplifying sample loading.
  • Syringe-barrel cartridges are compatible with vacuum manifolds and automated SPE devices.
  • Ultra-clean design prevents contamination and analytical interference.
  • Particle size of 60 μm is recommended for viscous samples.