Learn More
Waters Corp Oasis WAX Online Prospekt-2/Symbiosis Cartridge, 80Å, 30 µm, 10 mm, 96/pk
Catalog No. 50466023 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WAX cartridges for Symbiosis/Prospekt-2 systems are designed for rugged, reproducible, and ultra-fast on-line SPE in the Spark Holland Symbiosis and Prospekt-2 systems. Each cartridge contains 10.4 mg of Oasis WAX, a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. Each 800-series tray contains 96 cartridges and can be directly loaded into the Automatic Cartridge Exchange (ACE) or the ACE Feeder units of the Symbiosis/Prospekt-2 systems without the stainless steel tray holder/adaptor. For additional information on the Symbiosis and Prospekt-2 systems, go to http://www.spark.nl/
Catalog No. 50-466-023 Supplier Waters Corp Supplier No. 186004656
The Oasis WAX cartridges for Symbiosis/Prospekt-2 systems are designed for rugged, reproducible, and ultra-fast on-line SPE in the Spark Holland Symbiosis and Prospekt-2 systems. Each cartridge contains 10.4 mg of Oasis WAX, a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. Each 800-series tray contains 96 cartridges and can be directly loaded into the Automatic Cartridge Exchange (ACE) or the ACE Feeder units of the Symbiosis/Prospekt-2 systems without the stainless steel tray holder/adaptor. For additional information on the Symbiosis and Prospekt-2 systems, go to http://www.spark.nl/