Learn More
Waters Corp Waters Oasis WAX 96-well Plate, 10 mg Sorbent, 30 μm, Water Wettable
Catalog No. 50781159 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well plate features a polymeric reversed-phase, weak anion exchange mixed-mode sorbent for fast, simple, and highly selective sample preparation of strong acidic compounds. Its unique balance of hydrophobicity and water-wettability ensures consistent results even if wells dry out during critical steps. Compatible with various manifold configurations and most robotic liquid handling systems, it provides consistent results without evaporation or reconstitution, saving time.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • Designed for selective sample preparation of strong acidic compounds
  • Sorbent weight: 10 mg
  • Particle size: 30 μm
  • Ion exchange capacity: 0.6 meq/gram
  • Mass spectrometry compatible
  • Sorbent substrate: Copolymer
  • Water wettable for consistent results
  • pH range: 0 to 14
  • Material: Polypropylene
Catalog No. 50-781-159 Supplier Waters Corp Supplier No. 186002502
This 96-well plate features a polymeric reversed-phase, weak anion exchange mixed-mode sorbent for fast, simple, and highly selective sample preparation of strong acidic compounds. Its unique balance of hydrophobicity and water-wettability ensures consistent results even if wells dry out during critical steps. Compatible with various manifold configurations and most robotic liquid handling systems, it provides consistent results without evaporation or reconstitution, saving time.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent
  • Designed for selective sample preparation of strong acidic compounds
  • Sorbent weight: 10 mg
  • Particle size: 30 μm
  • Ion exchange capacity: 0.6 meq/gram
  • Mass spectrometry compatible
  • Sorbent substrate: Copolymer
  • Water wettable for consistent results
  • pH range: 0 to 14
  • Material: Polypropylene