Learn More
Waters Corp Waters 96-well Plate, Oasis WAX, 30 mg Sorbent per Well, 30 μm
Catalog No. 50781161 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WAX 96-well plate uses a polymeric reversed-phase, weak anion exchange mixed-mode sorbent, ideal for selective sample preparation of strong acidic compounds. It ensures consistent results by preventing issues if wells dry out, thanks to its balanced hydrophobicity and water-wettability. This plate is designed for use with diverse manifold configurations and robotic liquid handling systems.
  • Optimized for fast, simple, and selective sample preparation of strong acidic compounds.
  • Maintains performance with unique hydrophobicity and water-wettability, preventing issues if wells dry out.
  • Compatible with various manifold configurations and robotic liquid handling systems.
  • Improves recovery and reproducibility of solid phase extraction methods.
  • Offers pH stability, solvent compatibility, and exceptional polar chemical retention with a wide operational pH range of 0 to 14.
Catalog No. 50-781-161 Supplier Waters Corp Supplier No. 186002504
The Oasis WAX 96-well plate uses a polymeric reversed-phase, weak anion exchange mixed-mode sorbent, ideal for selective sample preparation of strong acidic compounds. It ensures consistent results by preventing issues if wells dry out, thanks to its balanced hydrophobicity and water-wettability. This plate is designed for use with diverse manifold configurations and robotic liquid handling systems.
  • Optimized for fast, simple, and selective sample preparation of strong acidic compounds.
  • Maintains performance with unique hydrophobicity and water-wettability, preventing issues if wells dry out.
  • Compatible with various manifold configurations and robotic liquid handling systems.
  • Improves recovery and reproducibility of solid phase extraction methods.
  • Offers pH stability, solvent compatibility, and exceptional polar chemical retention with a wide operational pH range of 0 to 14.