Learn More
Waters Corp Waters 96-well Plate, WAX Sorbent, 60 mg, 30 μm
Catalog No. 50782065 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WAX 96-well plate utilizes a polymeric reversed-phase, weak anion exchange mixed-mode sorbent, optimized for selective sample preparation of strong acidic compounds. Its unique balance of hydrophobicity and water-wettability prevents poor results if wells dry out. Compatible with various manifold configurations and robotic liquid handling systems.
  • Contains polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds.
  • Unique balance of hydrophobicity and water-wettability means results will not be poor if individual wells dry out during critical steps prior to sample loading.
  • Compatible with various manifold configurations and robotic liquid handling systems.
Catalog No. 50-782-065 Supplier Waters Corp Supplier No. 186003915
The Oasis WAX 96-well plate utilizes a polymeric reversed-phase, weak anion exchange mixed-mode sorbent, optimized for selective sample preparation of strong acidic compounds. Its unique balance of hydrophobicity and water-wettability prevents poor results if wells dry out. Compatible with various manifold configurations and robotic liquid handling systems.
  • Contains polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds.
  • Unique balance of hydrophobicity and water-wettability means results will not be poor if individual wells dry out during critical steps prior to sample loading.
  • Compatible with various manifold configurations and robotic liquid handling systems.