Learn More
Waters Corp Oasis μElution Plate, WAX, 96-well, 2 mg Sorbent, 30 μm
Catalog No. 50781157 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This patented 96-well WAX μElution plate is for SPE cleanup and analyte enrichment, featuring 2 mg of WAX sorbent per well. The polymeric reversed-phase, weak anion exchange mixed-mode sorbent is optimized for selective sample preparation of strong acidic compounds. Elution in 25 μL provides sensitive, robust, reproducible results without evaporation/reconstitution, compatible with automated high throughput SPE.
  • Designed for SPE cleanup and analyte enrichment (10 μL to 375 μL sample volumes).
  • Each well contains 2 mg of WAX sorbent.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent for selective preparation of strong acidic compounds.
  • Elution in 25 μL for sensitive, robust, reproducible results without evaporation/reconstitution.
  • 15-fold sample enrichment is possible.
  • Compatible with automated, high throughput SPE systems.
Catalog No. 50-781-157 Supplier Waters Corp Supplier No. 186002500
This patented 96-well WAX μElution plate is for SPE cleanup and analyte enrichment, featuring 2 mg of WAX sorbent per well. The polymeric reversed-phase, weak anion exchange mixed-mode sorbent is optimized for selective sample preparation of strong acidic compounds. Elution in 25 μL provides sensitive, robust, reproducible results without evaporation/reconstitution, compatible with automated high throughput SPE.
  • Designed for SPE cleanup and analyte enrichment (10 μL to 375 μL sample volumes).
  • Each well contains 2 mg of WAX sorbent.
  • Polymeric reversed-phase, weak anion exchange mixed-mode sorbent for selective preparation of strong acidic compounds.
  • Elution in 25 μL for sensitive, robust, reproducible results without evaporation/reconstitution.
  • 15-fold sample enrichment is possible.
  • Compatible with automated, high throughput SPE systems.