Learn More
Waters Corp Waters Oasis Vac Cartridge, WAX/GCB, 6cc, 200/50 mg, 60 μm
Catalog No. NC3314956 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is designed for environmental testing laboratories to perform low-level quantification of per- and polyfluoroalkyl substances (PFAS) in various environmental samples, including water, soil, and food. It features a combination of WAX and GCB in a single cartridge, streamlining the SPE process and saving time. Comprehensive quality control pre-screening for 32 PFAS prevents cross-contamination, ensuring accurate results and reducing false positives.
  • For sample preparation
  • WAX/GCB chemistry
  • Vac cartridge format
  • 0.6 meq/gram ion exchange capacity
  • Mass spec compatible
  • 60 μm particle size
  • 80 Å pore size
  • 200 mg WAX, 50 mg GCB sorbent weight
  • pH range: 0 to 14 pH
  • Polypropylene material
  • EPA Draft Method 1633 compatible
Catalog No. NC3314956 Supplier Waters Corp Supplier No. 186011111
This product is designed for environmental testing laboratories to perform low-level quantification of per- and polyfluoroalkyl substances (PFAS) in various environmental samples, including water, soil, and food. It features a combination of WAX and GCB in a single cartridge, streamlining the SPE process and saving time. Comprehensive quality control pre-screening for 32 PFAS prevents cross-contamination, ensuring accurate results and reducing false positives.
  • For sample preparation
  • WAX/GCB chemistry
  • Vac cartridge format
  • 0.6 meq/gram ion exchange capacity
  • Mass spec compatible
  • 60 μm particle size
  • 80 Å pore size
  • 200 mg WAX, 50 mg GCB sorbent weight
  • pH range: 0 to 14 pH
  • Polypropylene material
  • EPA Draft Method 1633 compatible