Learn More
Waters Corp Oasis WAX/GCB for PFAS Analysis 6cc Vac Cartridge, 200 mg WAX, 50 mg GCB, 60 µm WAX Particle Size, 30/pk;
Catalog No. NC3314955 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis WAX/GCB for PFAS Analysis 6cc Vac Cartridge, 200 mg WAX, 50 mg GCB, 60 µm WAX Particle Size, 30/pk;
Catalog No. NC3314955 Supplier Waters Corp Supplier No. 186011110
Oasis WAX/GCB for PFAS Analysis 6cc Vac Cartridge, 200 mg WAX, 50 mg GCB, 60 µm WAX Particle Size, 30/pk;