Learn More
Waters Corp Waters Solid Phase Extraction Cartridge, WCX, 225 mg Sorbent, 60 μm
Catalog No. 50179319 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Oasis WCX Plus cartridges feature a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for selective sample preparation of strong basic compounds and quaternary amines. The sorbent is water wettable, maintaining retention and recovery even if it runs dry. The 60 μm particle size is ideal for viscous samples, and the syringe-barrel design supports vacuum manifolds and automated SPE.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable sorbent maintains retention and excellent recoveries even if it runs dry.
  • 60 μm particle size for viscous samples.
  • Syringe-barrel design for vacuum manifolds and automated SPE instruments.
  • Broadest possible pH range of 0 to 14.
  • 0.75 meq/g of carboxylic-acid-ion-exchange capability.
  • Luer inlet hub and outlet tip.
Catalog No. 50-179-319 Supplier Waters Corp Supplier No. 186003518
These Oasis WCX Plus cartridges feature a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for selective sample preparation of strong basic compounds and quaternary amines. The sorbent is water wettable, maintaining retention and recovery even if it runs dry. The 60 μm particle size is ideal for viscous samples, and the syringe-barrel design supports vacuum manifolds and automated SPE.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable sorbent maintains retention and excellent recoveries even if it runs dry.
  • 60 μm particle size for viscous samples.
  • Syringe-barrel design for vacuum manifolds and automated SPE instruments.
  • Broadest possible pH range of 0 to 14.
  • 0.75 meq/g of carboxylic-acid-ion-exchange capability.
  • Luer inlet hub and outlet tip.