Learn More
Waters Corp Waters Vac Cartridge, 1 cc, 10 mg, 30 μm, WCX
Catalog No. 50201495 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1 cc cartridges are designed for solid phase extraction, featuring a polymeric reversed-phase, weak ion exchange mixed-mode sorbent. Optimized for fast and selective preparation of strong basic compounds and quaternary amines, they are water wettable, ensuring consistent performance even if the sorbent runs dry.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable sorbent maintains retention and recoveries if it runs dry.
  • Flangeless configuration enables close packing in automated SPE systems.
  • Contains 10 mg of high-quality sorbent.
  • Features a fine, 30-μm frit to reduce blockage and improve fluid flow.
  • Ensures high batch-to-batch and lot-to-lot reproducibility.
Catalog No. 50-201-495 Supplier Waters Corp Supplier No. 186004650
These 1 cc cartridges are designed for solid phase extraction, featuring a polymeric reversed-phase, weak ion exchange mixed-mode sorbent. Optimized for fast and selective preparation of strong basic compounds and quaternary amines, they are water wettable, ensuring consistent performance even if the sorbent runs dry.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable sorbent maintains retention and recoveries if it runs dry.
  • Flangeless configuration enables close packing in automated SPE systems.
  • Contains 10 mg of high-quality sorbent.
  • Features a fine, 30-μm frit to reduce blockage and improve fluid flow.
  • Ensures high batch-to-batch and lot-to-lot reproducibility.