Learn More
Waters Corp Waters Oasis WCX Cartridge, 1 cc/30 mg, 30 μm
Catalog No. 50781151 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Oasis WCX (Weak Cation eXchange) SPE material is designed for sample preparation of strong bases and quaternary amines. Its retention mechanism is mixed-mode (both ion exchange and reversed phase), which enhances retention for all types of basic analytes, particularly strong bases.
  • Designed for sample preparation of strong bases and quaternary amines.
  • Utilizes mixed-mode retention (ion exchange and reversed phase).
  • Enhances retention for all types of basic analytes.
  • Particularly effective for strong bases.
  • Format: 1 cc/30 mg.
  • Particle size: 30 μm.
  • Quantity: 100 per box.
Catalog No. 50-781-151 Supplier Waters Corp Supplier No. 186002494
The Oasis WCX (Weak Cation eXchange) SPE material is designed for sample preparation of strong bases and quaternary amines. Its retention mechanism is mixed-mode (both ion exchange and reversed phase), which enhances retention for all types of basic analytes, particularly strong bases.
  • Designed for sample preparation of strong bases and quaternary amines.
  • Utilizes mixed-mode retention (ion exchange and reversed phase).
  • Enhances retention for all types of basic analytes.
  • Particularly effective for strong bases.
  • Format: 1 cc/30 mg.
  • Particle size: 30 μm.
  • Quantity: 100 per box.