Learn More
Waters Corp Oasis Vac Cartridge, 3 cc, 60 mg Sorbent, 30 μm, WCX
Catalog No. 50781152 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 3 cc cartridges feature a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for highly selective sample preparation of strong basic compounds and quaternary amines. Its water wettable nature ensures higher retention and excellent recoveries, even if the sorbent runs dry. Designed for use with vacuum manifolds and automated SPE instruments, these cartridges offer a rugged solution for SPE methods across a wide pH range of 0 to 14.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable, maintaining high retention and excellent recoveries even if dry.
  • Syringe-barrel-type cartridges for vacuum manifolds and automated SPE instruments.
  • Stable from pH 0 to 14.
  • Delivers 0.75 meq/g of carboxylic acid ion-exchange capacity.
Catalog No. 50-781-152 Supplier Waters Corp Supplier No. 186002495
These 3 cc cartridges feature a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, optimized for highly selective sample preparation of strong basic compounds and quaternary amines. Its water wettable nature ensures higher retention and excellent recoveries, even if the sorbent runs dry. Designed for use with vacuum manifolds and automated SPE instruments, these cartridges offer a rugged solution for SPE methods across a wide pH range of 0 to 14.
  • Polymeric reversed-phase, weak ion exchange mixed-mode sorbent.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable, maintaining high retention and excellent recoveries even if dry.
  • Syringe-barrel-type cartridges for vacuum manifolds and automated SPE instruments.
  • Stable from pH 0 to 14.
  • Delivers 0.75 meq/g of carboxylic acid ion-exchange capacity.