Learn More
Waters Corp Waters Vac Cartridge, Oasis WCX, 3 cc, 60 mg, 60 μm
Catalog No. 50781154 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 3 cc Oasis WCX cartridges contain a polymeric reversed-phase, weak ion exchange mixed-mode sorbent for fast, simple, and highly selective sample preparation of strong basic compounds and quaternary amines. Its water-wettable nature ensures consistent retention and excellent recoveries even if the sorbent beds dry out. The 60 μm particle size is recommended for viscous samples.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water-wettable sorbent maintains high retention and excellent recoveries, even when dry.
  • 60 μm particle size is ideal for viscous samples.
  • Syringe-barrel cartridges are compatible with vacuum manifolds and automated SPE instruments.
  • Ensures high purity, consistency, and quality for sample analysis.
  • Delivers high recovery, reduced matrix effects, and unsurpassed selectivity.
Catalog No. 50-781-154 Supplier Waters Corp Supplier No. 186002497
The 3 cc Oasis WCX cartridges contain a polymeric reversed-phase, weak ion exchange mixed-mode sorbent for fast, simple, and highly selective sample preparation of strong basic compounds and quaternary amines. Its water-wettable nature ensures consistent retention and excellent recoveries even if the sorbent beds dry out. The 60 μm particle size is recommended for viscous samples.
  • Optimized for selective sample preparation of strong basic compounds and quaternary amines.
  • Water-wettable sorbent maintains high retention and excellent recoveries, even when dry.
  • 60 μm particle size is ideal for viscous samples.
  • Syringe-barrel cartridges are compatible with vacuum manifolds and automated SPE instruments.
  • Ensures high purity, consistency, and quality for sample analysis.
  • Delivers high recovery, reduced matrix effects, and unsurpassed selectivity.