Learn More
Waters Corp Waters Corporation Vac Cartridge, WCX, 150 mg Sorbent, 30 μm
Catalog No. 50781155 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vac cartridge contains a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, ideal for highly selective sample preparation of strong basic compounds and quaternary amines. It is water wettable, ensuring high retention and recoveries even if the sorbent runs dry, and is designed for use with vacuum manifolds and automated SPE instruments.
  • Leverages WCX (Weak Cation Exchange) SPE sorbent.
  • Optimized for fast and simple delivery.
  • Ion exchange capacity of 0.75 meq/g.
  • Particle size of 30 μm.
  • Mass spectrometry compatible.
  • Provides a wide and stable pH range (0 to 14).
  • Ultra-clean to ensure no contamination interferes with analysis or sample preparation.
Catalog No. 50-781-155 Supplier Waters Corp Supplier No. 186002498
This vac cartridge contains a polymeric reversed-phase, weak ion exchange mixed-mode sorbent, ideal for highly selective sample preparation of strong basic compounds and quaternary amines. It is water wettable, ensuring high retention and recoveries even if the sorbent runs dry, and is designed for use with vacuum manifolds and automated SPE instruments.
  • Leverages WCX (Weak Cation Exchange) SPE sorbent.
  • Optimized for fast and simple delivery.
  • Ion exchange capacity of 0.75 meq/g.
  • Particle size of 30 μm.
  • Mass spectrometry compatible.
  • Provides a wide and stable pH range (0 to 14).
  • Ultra-clean to ensure no contamination interferes with analysis or sample preparation.