Learn More
Waters Corp Waters Oasis WCX Cartridge, Mixed-Mode Weak Cation eXchange, 6 cc, 500 mg, 60 μm
Catalog No. 50466018 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Oasis WCX is a Mixed-Mode Weak Cation eXchange sorbent for strong bases and quaternary amines. It is stable from pH 0–14, water-wettable, and based on the Oasis HLB Copolymer, offering reversed-phase and ion exchange modes of retention for enhanced cleanup selectivity and sensitivity.
  • Provides both reversed-phase and ion exchange modes of retention.
  • Enables greater cleanup selectivity and sensitivity for acidic and/or basic compounds.
  • Built upon the unique water-wettable Oasis HLB Copolymer.
  • Allows for concentration of samples, especially with μElution Plate format (up to 15x concentration without evaporation).
  • Offers a variety of tools for method development to optimize protocols for maximum selectivity and sensitivity.
  • Helps achieve high recovery and reproducible results for a wide range of compounds.
Catalog No. 50-466-018 Supplier Waters Corp Supplier No. 186004646
Oasis WCX is a Mixed-Mode Weak Cation eXchange sorbent for strong bases and quaternary amines. It is stable from pH 0–14, water-wettable, and based on the Oasis HLB Copolymer, offering reversed-phase and ion exchange modes of retention for enhanced cleanup selectivity and sensitivity.
  • Provides both reversed-phase and ion exchange modes of retention.
  • Enables greater cleanup selectivity and sensitivity for acidic and/or basic compounds.
  • Built upon the unique water-wettable Oasis HLB Copolymer.
  • Allows for concentration of samples, especially with μElution Plate format (up to 15x concentration without evaporation).
  • Offers a variety of tools for method development to optimize protocols for maximum selectivity and sensitivity.
  • Helps achieve high recovery and reproducible results for a wide range of compounds.