Learn More
Waters Corp WATERS Oasis WCX 1 cc Flangeless Vac Cartridge, 10 mg, 30 μm
Catalog No. 50201607 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 1 cc flangeless cartridges feature a polymeric reversed-phase, weak ion exchange mixed-mode sorbent. Their water-wettable nature ensures consistent performance and excellent recoveries, even if the sorbent beds dry out before sample loading. The flangeless design allows for close packing in specialized automated solid phase extraction (SPE) systems.
  • Optimized for fast, simple, and highly selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable, maintaining higher retention and excellent recoveries.
  • Flangeless configuration for close packing in automated SPE systems.
  • Mass spectrometry compatible.
  • PH range of 0 to 14.
  • Ion exchange capacity of 0.75 meq/gram.
  • Sorbent substrate is copolymer.
  • Constructed from polypropylene material.
Catalog No. 50-201-607 Supplier Waters Corp Supplier No. 186006342
These 1 cc flangeless cartridges feature a polymeric reversed-phase, weak ion exchange mixed-mode sorbent. Their water-wettable nature ensures consistent performance and excellent recoveries, even if the sorbent beds dry out before sample loading. The flangeless design allows for close packing in specialized automated solid phase extraction (SPE) systems.
  • Optimized for fast, simple, and highly selective sample preparation of strong basic compounds and quaternary amines.
  • Water wettable, maintaining higher retention and excellent recoveries.
  • Flangeless configuration for close packing in automated SPE systems.
  • Mass spectrometry compatible.
  • PH range of 0 to 14.
  • Ion exchange capacity of 0.75 meq/gram.
  • Sorbent substrate is copolymer.
  • Constructed from polypropylene material.