Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Oasis WAX Flangeless Vac Cartridge, 3 cc, 60 mg Sorbent, 60 μm

Catalog No. 50201647 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These 3 cc flangeless cartridges contain a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds, the sorbent is water wettable, ensuring high retention and excellent recoveries even if it runs dry. The flangeless configuration enables close packing for automated solid phase extraction systems.

  • Contains a polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Barrel size: 3 cc.
  • Flangeless vac cartridge format.
  • Sorbent weight: 60 mg.
  • Particle size: 60 μm.
  • Ion exchange capacity: 0.75 meq/gram.
  • Mixed mode operation.
  • Pore size: 80 Å.
  • Copolymer sorbent substrate.
  • Water wettable.
  • pH range: 0 to 14.
  • Material: Polypropylene.
  • Units per package: 100.

Catalog No. 50-201-647 Supplier Waters Corp Supplier No. 186006502
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These 3 cc flangeless cartridges contain a polymeric reversed-phase, weak anion exchange mixed-mode sorbent. Optimized for fast, simple, and highly selective sample preparation of strong acidic compounds, the sorbent is water wettable, ensuring high retention and excellent recoveries even if it runs dry. The flangeless configuration enables close packing for automated solid phase extraction systems.

  • Contains a polymeric reversed-phase, weak anion exchange mixed-mode sorbent.
  • Barrel size: 3 cc.
  • Flangeless vac cartridge format.
  • Sorbent weight: 60 mg.
  • Particle size: 60 μm.
  • Ion exchange capacity: 0.75 meq/gram.
  • Mixed mode operation.
  • Pore size: 80 Å.
  • Copolymer sorbent substrate.
  • Water wettable.
  • pH range: 0 to 14.
  • Material: Polypropylene.
  • Units per package: 100.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.