Learn More
Waters Corp Waters Oasis Flangeless Vac Cartridge, WCX, 1 cc, 30 mg Sorbent, 30 μm
Catalog No. 50201644 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These flangeless vac cartridges feature Oasis WCX sorbent, a polymeric reversed-phase, weak ion exchange mixed-mode sorbent. They are optimized for fast, simple, and highly selective sample preparation of strong basic compounds and quaternary amines.
  • Sorbent is water wettable, maintaining high retention and excellent recoveries even if dry.
  • Eliminates the need for extraordinary steps to prevent sorbent beds from drying out during pre-sample loading.
  • Flangeless configuration allows cartridges to be closely packed for processing in specialized automated SPE systems.
Catalog No. 50-201-644 Supplier Waters Corp Supplier No. 186006499
Add to Cart
Add to Cart
These flangeless vac cartridges feature Oasis WCX sorbent, a polymeric reversed-phase, weak ion exchange mixed-mode sorbent. They are optimized for fast, simple, and highly selective sample preparation of strong basic compounds and quaternary amines.
  • Sorbent is water wettable, maintaining high retention and excellent recoveries even if dry.
  • Eliminates the need for extraordinary steps to prevent sorbent beds from drying out during pre-sample loading.
  • Flangeless configuration allows cartridges to be closely packed for processing in specialized automated SPE systems.